Learn More
Medchemexpress LLC Bragsin1 | 369631-68-5 | 99.91% | 273.16 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY111549100MG
Encompass_Preferred
Bragsin1 is a potent, selective, and noncompetitive inhibitor of the ArfGEF BRAG2, which inhibits Arf GTPase activation, with an IC50 of 3 μM. It binds to the PH domain of BRAG2 and acts as a noncompetitive interfacial inhibitor. Bragsin1 does not affect the Sec7 domain of human ArfGEFs and shows anti-cancer activity.
  • Potent, selective, and noncompetitive inhibitor of ArfGEF BRAG2.
  • Inhibits Arf GTPase activation with an IC50 of 3 μM.
  • Binds to the PH domain of BRAG2.
  • Acts as a noncompetitive interfacial inhibitor.
  • Does not affect the Sec7 domain of human ArfGEFs.
  • Shows anti-cancer activity.
Catalog No. 50-003-46739