Learn More
Medchemexpress LLC HY-111549 5mg Medchemexpress, Bragsin1 CAS:369631-68-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1115495MG
Encompass_Preferred
Medchemexpress, HY-111549 5mg Bragsin1 CAS:369631-68-5 Bragsin1 is a potent, selective and noncompetitive inhibitor of the ArfGEF BRAG2, inhibits Arf GTPase activation, with an IC50 of 3 μM. Bragsin1 binds to PH domain of BRAG2, and is a noncompetitive interfacial inhibitor. Bragsin1 has no effect on the Sec7 domain of human ArfGEFs. Anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8916