Learn More
Bioworld Brain-Heart Infusion Broth, 2 kg
Supplier:  Bioworld 306200133
Encompass_Preferred
Brain-Heart Infusion BrothUsed in the cultivation of a wide variety of fastidious microorganisms, including streptococci, pneumococci and meningococci from a variety of clinical and nonclincal sourcespH: 7.4Prepared Appearance: light to medium amber brothContains:Brain Heart Infusion from Solids Pancreatic Digest of Gelatin Sodium Chloride Dextrose Disodium Phosphate
Catalog No. 50-254-283