Learn More
Avanti Polar Lipids BRAIN SM 25MG
Supplier:  Avanti Polar Lipids 860062P25MG
Encompass
Brain SM, Sphingomyelin (Brain, Porcine), Purity: > 99%, Molecular Weight: 731.09, Molecular Formula: C 41 H 83 N2O6P, Storage: -20 deg.C, Shelf Life: 2 Years, Powder form, White Color, CAS No: 85187-10-6, 25mg
Catalog No. NC9132761