Learn More
Medchemexpress LLC Branaplam hydrochloride | 1562338-39-9 | 99.4% | 429.94 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY19620A5MG
Encompass_Preferred
Branaplam hydrochloride is a highly potent, selective, and orally active survival motor neuron-2 (SMN2) splicing modulator. It is for research use only, inhibiting human-ether-a-go-go-related gene (hERG) and showing potential in treating spinal muscular atrophy.
  • Highly potent, selective, and orally active splicing modulator.
  • Inhibits human-ether-a-go-go-related gene (hERG).
  • Elevates full-length SMN protein.
  • Extends survival in a severe spinal muscular atrophy (SMA) mouse model.
Catalog No. 50-004-00911