Learn More
Medchemexpress LLC Branaplam hydrochloride | 1562338-39-9 | MFCD29472288 | 99.4% | 429.94 g/mol | C22H27N5O2·HCl | 10 MG
SDP
Supplier:  Medchemexpress LLC HY19620A10MG
Encompass_Preferred
Branaplam hydrochloride is an orally active small-molecule modulator of SMN2 pre-mRNA splicing that elevates full-length survival motor neuron (SMN) protein in preclinical models. It has been investigated in neurodegenerative disease research and is supplied for laboratory research use.
  • Orally active small-molecule splicing modulator.
  • Potent activity with reported EC50 ≈ 20 nM for SMN.
  • Elevates full-length SMN protein in preclinical models.
  • Suitable for in vitro and in vivo research studies.
  • High chemical purity (>99%).
  • Provided as hydrochloride salt for improved handling and solubility.
Catalog No. 50-002-99064