Learn More
Selleck Chemical LLC Branebrutinib S8832-5MG
SDP
Supplier:  Selleck Chemical LLC S88325MG
Encompass_Preferred
Branebrutinib (BMS-986195) is a potent inhibitor of BTK with IC50 values of 0 1 nM 0 9 nM 1 5 nM 5 nM for BTK TEC BMX TXK respectively
Catalog No. 50-000-25382