Learn More
Cayman Chemical BrasIcasterol 10mg
SDP
Supplier:  Cayman Chemical 2229610 MG
Encompass_Preferred
A phytosterol; inhibits recombinant sterol Δ24-reductase (Ki = 42.7 μM) in vitro; increases accumulation of desmosterol and inhibits cholesterol formation in HL-60 cells in a dose-dependent manner; inhibits the hemolytic activity of pneumolysin from S. pneumoniae
Catalog No. 50-296-4340