Learn More
New Jersey Fluid Systems LLC BRASS 1 PIECE 40 SER BALL VALV
Supplier:  New Jersey Fluid Systems LLC B41S2
Encompass
Brass Swagelok 40 in. series Ball valve. 1/8 in. OD Tube Fitting Ends. 2,500 psi @100F +50F to 150F PTFE Packing.
Catalog No. NC0378943