Learn More
Perkin Elmer US LLC Brass Long Term Storage Cap
Supplier:  Perkin Elmer US LLC 09908851
Encompass_Preferred
Two end caps required per tube. Also requires PTFE ferrule (04964467).
Catalog No. 50-211-3089