Learn More
Medchemexpress LLC Brassinazole | 224047-41-0 | >=98.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC HY12116125MG
Encompass_Preferred
Brassinazole is a triazole-type compound that inhibits brassinosteroid (BR) biosynthesis by increasing inhibition of CYP90B. It is supplied for research use and is provided as solid samples and as a 10 mM solution in DMSO for experimental use.
  • Triazole-type brassinosteroid biosynthesis inhibitor.
  • Increases inhibition of CYP90B.
  • Used to probe brassinosteroid and gibberellin biosynthesis pathways in plant research.
  • Available as multiple solid pack sizes and as a 10 mM DMSO solution.
  • Molecular formula C18H18ClN3O and molecular weight ~327.81 g/mol.
Catalog No. 50-002-33499