Learn More
Medchemexpress LLC Brassinazole | 224047-41-0 | ≥98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12116150MG
Encompass_Preferred
(Rac)-Brassinazole is a triazole-type small molecule that inhibits brassinosteroid (BR) biosynthesis by increasing inhibition of CYP90B. It is used as a research reagent to study plant hormone pathways, including reported effects on gibberellin biosynthesis.
  • Triazole-type inhibitor of brassinosteroid biosynthesis.
  • Increases inhibition of CYP90B in BR biosynthesis.
  • Applicable for research on plant growth and hormone interactions, including gibberellin pathways.
  • Available in solid and prepared DMSO solution formats for assay flexibility.
  • Recommended storage: powder at -20°C (long-term) or 4°C (short-term); in solvent at -80°C or -20°C.
Catalog No. 50-002-33935