Learn More
Medchemexpress LLC HY-N0273 5mg Medchemexpress, Brassinolide CAS:72962-43-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN02735MG
Encompass_Preferred
Medchemexpress, HY-N0273 5mg Brassinolide CAS:72962-43-7 Brassinolide is a predominant plant growth modulator that regulate plant cell elongation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3503