Learn More
Medchemexpress LLC Brazilin | 474-07-7 | 99.2% | 286.28 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN007250MG
Encompass_Preferred
Brazilin is a red dye precursor obtained from the heartwood of several species of tropical hardwoods. It exhibits various biological activities, including inhibition of cell proliferation, promotion of apoptosis, and induction of autophagy.
  • Inhibits cell proliferation and promotes apoptosis.
  • Induces autophagy through the AMPK/mTOR pathway.
  • Demonstrates chondroprotective activities.
  • Exhibits anti-inflammatory activities.
Catalog No. 50-004-30186