Learn More
Medchemexpress LLC BRD-K98645985 10mM 1mL | 1357647-78-9 | 1 ML

Supplier: Medchemexpress LLC HY11426810MM 1ML
BRD-K98645985 is a small-molecule inhibitor of BAF (mammalian SWI/SNF) complexes that reverses HIV-1 latency in vitro. It is supplied as a 10 mM solution in DMSO (1 mL) and is also available as solid quantities for research use.
- Ready-to-use 10 mM solution in DMSO, 1 mL.
- Available as solid quantities: 1 mg, 5 mg, 10 mg, larger sizes by request.
- High purity by HPLC (reported 98.67%).
- Molecular weight 573.73 and formula C33H43N5O4.
- Storage: powder -20°C (long term) or 4°C (short term); in solvent -80°C or -20°C per stability data.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.