Learn More
Medchemexpress LLC BRD0539 | 1403838-79-8 | 98.33% | 452.54 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY136251100MG
Encompass_Preferred
BRD0539 is a Streptococcus pyogenes Cas9 (SpCas9) inhibitor with an IC50 of 22 μM in an in vitro DNA cleavage assay.
  • Dose-dependently blocks the formation of the DNA-bound state.
  • Impairs the perturbation induced by 4PAM DNA.
  • Does not interfere with the SpCas9:gRNA interaction.
  • Inhibits SpCas9 in the eGFP-disruption assay.
  • Unable to inhibit FnCpf1.
Catalog No. 50-003-49708