Learn More
Selleck Chemical LLC BRD0705 S9638-5MG
SDP
Supplier:  Selleck Chemical LLC S96385MG
Encompass_Preferred
BRD0705 is a potent paralog selective and orally active inhibitor of GSK3 (Glycogen synthase kinase 3 ) with IC50 of 66 nM and Kd of 4 8 M BRD0705 also inhibits GSK3 with IC50 of 515 nM BRD0705 can be used for acute myeloid leukemia (AML)
Catalog No. 50-000-24250