Learn More
Cayman Chemical BRD4 bromodomaIn 1humnre 100ug
SDP
Supplier:  Cayman Chemical 11720100 UG
Encompass_Preferred
Source: Recombinant N-terminal His-tagged BRD4 bromodomain 1 expressed in E. coli • Amino acids: 49-170 (partial protein) • MW: 16.6 kDa
Catalog No. 50-295-2310