Learn More
Cayman Chemical BRD4 bromodomaIn 1humnre 100ug
SDP
Supplier:  Cayman Chemical 11068100 UG
Encompass_Preferred
Source: Recombinant human N-terminal GST-tagged BRD4 bromodomain 1 expressed in E. coli • Amino acids: 49-170 • MW: 41.4 kDa
Catalog No. 50-295-1398