Learn More
Cayman Chemical BRD4 bromodomaIn 2humnre 100ug
SDP
Supplier:  Cayman Chemical 11066100 UG
Encompass_Preferred
Source: Recombinant N-terminal GST-tagged BRD4 bromodomain 2 expressed in E. coli • Amino acids: 342-460 • MW: 40.6 kDa
Catalog No. 50-295-1129