Learn More
TARGETMOL CHEMICALS INC BRD4097 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T305795MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BRD4097 a negative control in the HDAC1/2/3/8 assay is a selective histone deacetylase (HDAC3) inhibitor that can be used to protect b-cells and improve insulin resistance help promote cognitive function and enhance learning and memory formation. purity: 99%
Catalog No. 50-258-0405