Learn More
Selleck Chemical LLC BrdU
SDP
Supplier:  Selleck Chemical LLC S7918500MG
Encompass_Preferred
BrdU (Bromodeoxyuridine) is a nucleoside analog that competes with thymidine for incorporation into DNA and used in the detection of proliferating cells
Catalog No. 50-000-00561