Learn More
MilliporeSigma BRDU IHC DETECTION KIT
Supplier:  MilliporeSigma 2760
Encompass
BrdU IHC Kit, Contains 4X Trypsin enzyme Conc, Trypsin dilu soln, Denaturing soln, Blocking buffer, Detector antibody, Streptavidin-HRP Conjugate, Substrate reactn buffer, DAB conc, Hematoxylin counterstain, Mounting Media etc, 50 slides
Catalog No. 50-818-497