Learn More
Amsbio LLC Breast cancer tissue array (mo
Supplier:  Amsbio LLC BR1202A
Encompass
Breast cancer tissue array (most of triple negative TNBC) including pathology grade TNM clinical stage and IHC marker (ER PR and Her-2) 60 cases/120 cores(core size 1.5mm) replacing BR1202
Catalog No. 50-256-4541