Learn More
Cayman Chemical BrefeldIn A 10mg
SDP
Supplier:  Cayman Chemical 1186110 MG
Encompass_Preferred
A natural fungal metabolite which reversibly interferes with protein trafficking and secretion mediated by the Golgi apparatus and endoplasmic reticulum; directly and reversibly inhibits Sec7 domain-containing GEFs which are necessary for Arf activation associated with vesicular transport (IC50 = ~10 μM)
Catalog No. 50-295-2866