Learn More
Selleck Chemical LLC Brefeldin A
SDP
Supplier:  Selleck Chemical LLC S70461G
Encompass_Preferred
Brefeldin A (BFA) is a lactone antibiotic and ATPase inhibitor for protein transport with IC50 of 0 2 M in HCT 116 cells induces cancer cell differentiation and apoptosis It could also improve the HDR(homology-directed repair) efficiency and be an enhancer of CRISPR-mediated HDR Brefeldin A is also an inhibitor of autophagy and mitophagy
Catalog No. 50-000-01827