Learn More
Selleck Chemical LLC Brepocitinib S8804-1G
SDP
Supplier:  Selleck Chemical LLC S88041G
Encompass_Preferred
Brepocitinib (PF-06700841 PF-841) is a potent inhibitor of Tyk2 and Jak1 with IC50s of 23 nM 17 nM 77 nM for Tyk2 Jak1 and Jak2 respectively It has appropriate in-family selectivity against JAK2 and JAK3
Catalog No. 50-000-24246