Learn More
TARGETMOL CHEMICALS INC Brepocitinib P-Tosylate 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1242710MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.Brepocitinib P-Tosylate (PF-06700841 P-Tosylate) is a potent dual inhibitor of Janus kinase 1 (JAK1) and TYK2 (IC50s of 17 nM and 23 nM, respectively). Purity 98.99%
Catalog No. 50-287-8389