Learn More
Medchemexpress LLC Brequinar | 96187-53-0 | MFCD00866437 | 99.9% | 375.37 | C23H15F2NO2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10832525MG
Encompass_Preferred
Brequinar is a research-grade small-molecule inhibitor of dihydroorotate dehydrogenase (DHODH) used in biochemical and pharmacological studies, including antiviral and antineoplastic research. Identifiers: CAS 96187-53-0; formula C23H15F2NO2; molecular weight 375.37. Supplied as a powder with recommended storage at -20°C for long-term stability.
  • Potent dihydroorotate dehydrogenase (DHODH) inhibitor with low-nanomolar activity.
  • High reported purity (99.9%) suitable for biological testing.
  • Supplied as a powder for convenient handling and dissolution.
  • Stable under recommended storage conditions for extended shelf life.
  • Appropriate for in vitro biochemical and cell-based assays.
Catalog No. 50-002-43330