Learn More
Selleck Chemical LLC Brequinar Sodium 5mg
Supplier: Selleck Chemical LLC S35655MG
Brequinar Sodium (Bipenquinate, BRQ, DUP-785, NSC 368390) is a potent and selective dihydroorotate dehydrogenase (DHODH) inhibitor with IC50 of ~20 nM and triggers differentiation in the ER-HoxA9, U937, and THP1 cells with ED50 of ~1 ?M. *For Research & Development use only. Product is not intended for drug, household, or human consumption.