Learn More
Selleck Chemical LLC Brequinar Sodium S3565-1g
SDP
Supplier:  Selleck Chemical LLC S35651G
Encompass_Preferred
Brequinar Sodium (Bipenquinate BRQ DUP-785 NSC 368390) is a potent and selective dihydroorotate dehydrogenase (DHODH) inhibitor with IC50 of 20 nM and triggers differentiation in the ER-HoxA9 U937 and THP1 cells with ED50 of 1 M
Catalog No. 50-310-9338