Learn More
Medchemexpress LLC Bretisilocin | 2698331-35-8 | 99.1% | 220.29 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY172420
Encompass_Preferred
Bretisilocin (GM-2505) is an agonist for the 5-HT2A receptor and a serotonin releaser. It exhibits antidepressant activity and can be used in the study of depression.
  • Agonist for 5-HT2A receptor and a serotonin releaser
  • Exhibits antidepressant activity
  • Good safety profile and dose-proportional pharmacokinetics
  • Appearance: oil
  • Color: white to light yellow
  • For research use only
Catalog No. 50-003-85614