Learn More
Medchemexpress LLC Bretisilocin | 2698331-35-8 | 99.7% | 220.29 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1724205MG
Encompass_Preferred
Bretisilocin (GM-2505) is an agonist for the 5-HT2A receptor and a serotonin releaser. It exhibits antidepressant activity and can be used in the study of depression. It has a good safety profile and exhibits dose-proportional pharmacokinetics.
  • Agonist for 5-HT2A receptor
  • Serotonin releaser
  • Exhibits antidepressant activity
  • Used in the study of depression
  • Good safety profile
  • Exhibits dose-proportional pharmacokinetics
Catalog No. 50-003-85039