Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Brevianamide F (cyclo(L-Pro-L-Trp)) | 38136-70-8 | MFCD29917326 | 99.0% | 283.32 g/mol | C16H17N3O2 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY10038510MM 1ML

Encompass_Preferred

Brevianamide F (Cyclo(L-Pro-L-Trp)) is an indole alkaloid and mycotoxin used in biochemical and pharmacological research. It is supplied as solid material in multiple mass sizes and as a ready-to-use 10 mM solution in DMSO (1 mL), suitable for assay development, screening, and analytical characterization.

  • High listed purity: 99.03%.
  • Identifying CAS number: 38136-70-8.
  • Chemical formula C16H17N3O2 and molecular weight 283.32 g/mol.
  • Available as solids in multiple sizes and as solution for immediate use.
  • Solvent: DMSO for the 10 mM, 1 mL solution.

Catalog No. 50-002-13982


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.