Learn More
Medchemexpress LLC Briakinumab 10mg | 10mg
SDP
Supplier:  Medchemexpress LLC HYP9948610MG
Encompass_Preferred
Briakinumab (ABT-874) is a fully human anti-IL-12/23p40 monoclonal antibody Briakinumab targets and neutralizes IL-12 and IL-23 Briakinumab can be used for the research of rheumatoid arthritis inflammatory bowel disease and multiple sclerosis[1]
Catalog No. 50-003-23576