Learn More
Selleck Chemical LLC Brigatinib (AP26113) 5mg 1197953-54-0
SDP
Supplier:  Selleck Chemical LLC S82295MG
Encompass_Preferred
Brigatinib (AP26113) is a potent and selective ALK (IC50, 0.6 nM) and ROS1 (IC50, 0.9 nM) inhibitor. It also inhibits IGF-1R, FLT3, and mutant variants of FLT3 (D835Y) and EGFR with lower potentcy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5822