Learn More
Electron Microscopy Sciences Compound Brightfield Microscope Binocular, Achromatic
SDP
Supplier:  Electron Microscopy Sciences CP102
Encompass_Preferred
Designed for ease of use, the CP Series allows a novice to approach the microscope and quickly analyze their specimen. Void of complicated features, the CP Series offers pre-centered lenses and condensers, preventing operator confusion.
High eye point eye pieces, a rear facing turret for easy slide handling and a choice of lenses that provide large, flat fields of view offer brilliant images and outstanding resolution.
Built to take rough handling, the CP Series' frame, focusing and stage mechanisms are all metal construction because plastic wears over time. Spring loaded objectives protect against abuse but a focus stop control prohibits stage ramming accidents. Anti fungal protection and all glass light collectors ensure these microscopes will outlast their industry leading 5 Year Warranty.
Type: Achromatic
Catalog No. 50-343-51