Learn More
Alkali Scientific BrightStar™ PVDF Membrane, Pore Size 0.22μm, 300 x 3000 mm, 1 Roll
SDP
Supplier:  Alkali Scientific XR730
Encompass
BrightStar™ 0.22µm Pore Size PVDF Membrane is used for protein sequencing and the transfer of proteins from SDS-PAGE gels to membrane. Compared to nitrocellulose, PVDF is easier to handle and stain, has increased solvent resistance, and higher signal-to-noise ratio. BrightStar 0.22µm Pore Size PVDF Membrane comes as a dual tissue paper protected (top and bottom), 300 x 3000 mm (30cm wide x 3.0 meter) roll. Comparative to nitrocellulose, PVDF generally has improved handling characteristics and staining capabilities, increased solvent resistance, and higher signal-to-noise ratio.
Catalog No. 50-233-5973