Learn More
Bioworld BRIJ®-35, 10%, 100 mL
Supplier:  Bioworld 402300041
Encompass_Preferred
Peroxide free detergent in single use ampoules. Ideal for protein isolation and sample preparation.Specification:Proteomics GradePeroxide content: <1ug/ml.Packaged as 10 mL Vials This product is sold for research use onlyAppearance: LiquidBoiling Point : 100 °C(lit.)Density : 0.99 g/mL±0.002 g/mL at 20 °CHS Code : 34021300Melting Point: 41-45 °C (lit.)RTECS Number: JR5990000
Catalog No. 50-255-643