Learn More
Crescent Chemical Co Inc BRIJ 35 PRACT.
SDP
Supplier:  Crescent Chemical Co Inc SE15230.01
Encompass_Preferred
Brij 35 pract. n ca. 23. HLB 16.9. Suitable for the isolation of functional membrane complexes (1). Also applied in chemiluminescence analysis (2, 3). Size - 100G Storage Conditions - +15 C TO +30 C Catalog Number - 15230.01
Catalog No. 50-247-365