Learn More
Medchemexpress LLC Brilanestrant | 1365888-06-7 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY12864100MG
Encompass_Preferred
Brilanestrant is an orally bioavailable selective estrogen receptor degrader (SERD) with an IC50 of 0.7 nM. It is intended for research use only.
  • Orally bioavailable selective estrogen receptor degrader
  • Potent ER-α binder
  • Full transcriptional antagonist with no agonism
  • Induces ER-α degradation
  • Shows efficacy in MCF-7 breast cancer cell viability assays
  • Exhibits low clearance and good oral bioavailability
  • Demonstrates tumor-growth inhibition in MCF-7 xenograft models
Catalog No. 50-003-42035