Learn More
Medchemexpress LLC Brilanestrant | 1365888-06-7 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY12864
Encompass_Preferred
Brilanestrant is an orally bioavailable selective estrogen receptor degrader (SERD) with an IC₅₀ of 0.7 nM. It demonstrates potency and efficacy in ER-α degradation and MCF-7 breast cancer cell viability assays, acting as a full transcriptional antagonist without agonism. It induces a distinct ERα conformation and does not exhibit tamoxifen-like ER agonism in MCF7 cells.
  • Potent ER-α binder.
  • Full transcriptional antagonist with no agonism.
  • Effective in ER-α degradation and MCF-7 breast cancer cell viability assays.
  • Induces a distinct ERα conformation and lacks tamoxifen-like ER agonism.
  • Exhibits low clearance and good bioavailability across species.
  • Demonstrates substantial tumor-growth inhibition in tamoxifen-sensitive MCF-7 xenograft models.
Catalog No. 50-003-42982