Learn More
Medchemexpress LLC Brilanestrant | 1365888-06-7 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1286450MG
Encompass_Preferred
Brilanestrant is an orally bioavailable selective estrogen receptor degrader (SERD) with an IC50 of 0.7 nM. It acts as a potent ER-α binder (IC50=6.1 nM) and a full transcriptional antagonist with no agonism (3x ERE, IC50=2 nM). It demonstrates good potency and efficacy in ER-α degradation (EC50=0.7 nM) and MCF-7 breast cancer cell viability (IC50=2.5 nM) assays.
  • Orally bioavailable selective estrogen receptor degrader (SERD)
  • Potent ER-α binder (IC50=6.1 nM)
  • Full transcriptional antagonist with no agonism
  • Induces a distinct ERα conformation
  • Does not show tamoxifen-like ER agonism in MCF7 cells
  • Low clearance and good bioavailability (40%-60%)
  • Substantial tumor-growth inhibition in tamoxifen-sensitive MCF-7 xenograft model
  • Displays dose-dependent efficacy in MCF7 xenograft model
Catalog No. 50-003-42566