Learn More
Medchemexpress LLC Brilliant Black BN | 2519-30-4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12838210MM/1ML
Encompass_Preferred
Brilliant Black BN (E151) is an azo dye and food colorant. It acts as a promising antiviral agent against EV71 infection by inhibiting the interaction between EV71 and its cellular uncoating factor cyclophilin A, suggesting its potential for investigating contagious diseases.
  • Functions as an azo dye and food colorant
  • Exhibits antiviral activity against EV71 infection
  • Inhibits EV71 interaction with cellular uncoating factor cyclophilin A
  • Useful for investigating contagious diseases
  • Effective against various EV71, CVA16, and CVA6 strains
  • Demonstrates a protective effect against EV71 in animal models
Catalog No. 50-003-39242