Learn More
Research Products International Corp Brilliant Blue R-250 250 G
SDP
Supplier:  Research Products International Corp B43000250.0
Encompass_Preferred
Brilliant Blue R-250 is commonly used for staining proteins in polyacrylamide gels.
Catalog No. 50-197-7294