Learn More
ION BIOSCIENCES Brilliant Thallium Snapshot Assay pIONeer kit - 2 plates
Supplier:  ION BIOSCIENCES 110112
Encompass_Preferred
ION Biosciences' Brilliant Thallium Snapshot Assay pIONeer kit is an endpoint-compatible potassium channel assay. Brilliant Thallium Snapshot kit is a thallium flux assay adaptation designed to work with standard plate readers, microscopes, flow cytometers, and more, for measuring flux through potassium (K⁺), sodium (Na⁺), and non-selective cation channels, transporters, and their effectors, including GPCRs. 2 plate kit.
Catalog No. NC4043238