Learn More
Biolegend Brilliant Violet 421TM anti-hu
Supplier:  Biolegend 359117
Encompass_Preferred
Brilliant Violet 421™ anti-human CD195 (CCR5) [J418F1]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2141