Learn More
Biolegend Brilliant Violet 650TM anti-hu
Supplier:  Biolegend 369648
Encompass_Preferred
Brilliant Violet 650™ anti-human CD152 (CTLA-4) [BNI3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests
Catalog No. 50-272-2083