Learn More
Selleck Chemical LLC Brimonidine Tartrate 50mg 70359-46-5
SDP
Supplier:  Selleck Chemical LLC S426650MG
Encompass_Preferred
Brimonidine Tartrate is a highly selective ?-adrenergic receptor agonist with EC50 of 0.45 nM for the ?2A adrenoreceptor, and used to treat open-angle glaucoma or ocular hypertension. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3821