Learn More
Medchemexpress LLC Brinzolamide | 138890-62-7 | 99.91% | 1 ML
Supplier: Medchemexpress LLC HYB058810MM/1ML
Brinzolamide (AL-4862) is a highly selective carbonic anhydrase II inhibitor, demonstrating an IC50 value of 3.2 nM. It functions by reducing intraocular pressure through the inhibition of ciliary CA-II, thereby decreasing atrial fluid secretion. This compound is utilized in the topical treatment of elevated intraocular pressure in individuals diagnosed with ocular hypertension or open-angle glaucoma.
- Selective carbonic anhydrase II inhibitor
- IC50 value of 3.2 nM
- Reduces intraocular pressure
- Inhibits ciliary CA-II
- Decreases atrial fluid secretion
- Used for ocular hypertension
- Used for open-angle glaucoma